Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP1
Entrez Gene ID
702710
Gene Name
Sp1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.11: 52949866 - 52986395 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151801.2 XP_015007287.1 - 120 122
GenBank mRNA JU336057.1 - - 96 122
JU476915.1 - - 86 121

View Full Product Details