Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN2
Entrez Gene ID
574368
Gene Name
neuroligin 2
Gene Aliases
-
Chromosome Location
Chr.16: 7171714 - 7186856 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278449.1 NP_001265378.1 - 737 75
XM_015118450.2 XP_014973936.1 - 1629 75
XM_028835458.1 XP_028691291.1 - 1551 75
XM_028835459.1 XP_028691292.1 - 1244 75
GenBank mRNA JV552452.1 - - 576 75
JV623614.1 - - 598 75
JV691211.1 - - 661 75
JV719711.1 - - 596 75

View Full Product Details