Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRTC2
Entrez Gene ID
715908
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
EGK_01342
Chromosome Location
Chr.1: 97103264 - 97114446 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261373.1 NP_001248302.1 - 684 88
XM_015152564.2 XP_015008050.1 - 746 103
XM_015152571.2 XP_015008057.1 - 677 103
XM_015152597.2 XP_015008083.1 - 1008 103
GenBank mRNA JU333300.1 - - 640 88
JU474613.1 - - 681 88
JV046393.1 - - 684 88

View Full Product Details