Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPRSS11A
Entrez Gene ID
711430
Gene Name
transmembrane serine protease 11A
Gene Aliases
-
Chromosome Location
Chr.5: 65585859 - 65650273 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028848176.1 XP_028704009.1 - 648 121
XM_028848177.1 XP_028704010.1 - 639 121

View Full Product Details