Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCMT1
Entrez Gene ID
702050
Gene Name
leucine carboxyl methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.20: 23464190 - 23536768 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260979.1 NP_001247908.1 - 848 78
XM_028840444.1 XP_028696277.1 - 876 78
XM_028840445.1 XP_028696278.1 - 876 78
XM_028840446.1 XP_028696279.1 - 876 78
XM_028840447.1 XP_028696280.1 - 876 78
GenBank mRNA JU321125.1 - - 845 78
JU473282.1 - - 719 78
JV047305.1 - - 848 78
JV653365.1 - - 851 78

View Full Product Details