Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCMT1
Entrez Gene ID
702050
Gene Name
leucine carboxyl methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.20: 23464190 - 23536768 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260979.1 NP_001247908.1 - 1037 91
XM_028840444.1 XP_028696277.1 - 1065 91
XM_028840445.1 XP_028696278.1 - 1065 91
XM_028840446.1 XP_028696279.1 - 1065 91
GenBank mRNA JU321125.1 - - 1034 91
JU473282.1 - - 908 91
JV047305.1 - - 1037 91
JV653365.1 - - 1040 91

View Full Product Details