Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF1
Entrez Gene ID
711908
Gene Name
transmembrane 4 L six family member 1
Gene Aliases
-
Chromosome Location
Chr.2: 51494372 - 51525824 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266715.1 NP_001253644.1 - 528 67
XM_015131757.2 XP_014987243.1 - 1255 67
GenBank mRNA JU322506.1 - - 520 67
JU473727.1 - - 528 67

View Full Product Details