Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRGN
Entrez Gene ID
712299
Gene Name
serglycin
Gene Aliases
PRG1
Chromosome Location
Chr.9: 68701782 - 68715355 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261094.2 NP_001248023.1 - 260 70
GenBank mRNA JU335343.1 - - 291 70
JU476295.1 - - 311 70
JV047576.1 - - 262 70
JV718293.1 - - 300 70

View Full Product Details