Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM35A
Entrez Gene ID
702205
Gene Name
transmembrane protein 35A
Gene Aliases
EGK_20734
Chromosome Location
Chr.X: 97882712 - 97898880 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127811.2 XP_014983297.1 - 1720 125

View Full Product Details