Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCRT2
Entrez Gene ID
716940
Gene Name
scratch family transcriptional repressor 2
Gene Aliases
-
Chromosome Location
Chr.10: 65875737 - 65891025 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108022.4 XP_001108022.3 - 1503 110

View Full Product Details