Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OXER1
Entrez Gene ID
713039
Gene Name
oxoeicosanoid receptor 1
Gene Aliases
EGK_05277
Chromosome Location
Chr.13: 66020683 - 66024757 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001110986.4 XP_001110986.2 - 3925 96

View Full Product Details