Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9H10orf90
Entrez Gene ID
696655
Gene Name
chromosome 9 C10orf90 homolog
Gene Aliases
-
Chromosome Location
Chr.9: 126867272 - 127105010 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088646.4 XP_001088646.2 - 184 142

View Full Product Details