Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9H10orf90
Entrez Gene ID
696655
Gene Name
chromosome 9 C10orf90 homolog
Gene Aliases
-
Chromosome Location
Chr.9: 126867272 - 127105010 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001088646.4 XP_001088646.2 - 1888 96
XM_015148444.2 XP_015003930.2 - 2441 96
XM_015148445.2 XP_015003931.2 - 1775 96
XM_028826994.1 XP_028682827.1 - 2623 96
XM_028826995.1 XP_028682828.1 - 1751 96
XM_028826996.1 XP_028682829.1 - 1830 96
GenBank mRNA JU325322.1 - - 471 96
JV706534.1 - - 195 96

View Full Product Details