Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO44
Entrez Gene ID
714585
Gene Name
F-box protein 44
Gene Aliases
-
Chromosome Location
Chr.1: 212798138 - 212807538 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260876.1 NP_001247805.1 - 820 114
XM_015151844.2 XP_015007330.1 - 566 114
XM_015151906.2 XP_015007392.1 - 614 114
GenBank mRNA JU319733.1 - - 820 114

View Full Product Details