Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKR1E2
Entrez Gene ID
711035
Gene Name
aldo-keto reductase family 1 member E2
Gene Aliases
-
Chromosome Location
Chr.9: 6019171 - 6039594 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194235.2 NP_001181164.1 - 971 141
XM_015146359.2 XP_015001845.1 - 1286 141
XM_015146360.2 XP_015001846.1 - 1171 141
XM_015146362.2 XP_015001848.1 - 1118 141
XM_015146363.2 XP_015001849.1 - 888 141
XM_028853468.1 XP_028709301.1 - 973 141
XM_028853469.1 XP_028709302.1 - 860 141
GenBank mRNA JU325905.1 - - 471 141

View Full Product Details