Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHI3L1
Entrez Gene ID
703401
Gene Name
chitinase 3 like 1
Gene Aliases
-
Chromosome Location
Chr.1: 60679271 - 60687110 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265920.1 NP_001252849.1 - 386 72
GenBank mRNA JU323002.1 - - 386 72
JV043861.1 - - 400 72
JV704305.1 - - 407 72

View Full Product Details