Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT16
Entrez Gene ID
705114
Gene Name
synaptotagmin 16
Gene Aliases
-
Chromosome Location
Chr.7: 123943761 - 124199433 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028851597.1 XP_028707430.1 - 2816 77
GenBank mRNA JU336635.1 - - 1761 77
JV724730.1 - - 1106 77

View Full Product Details