Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RERG
Entrez Gene ID
700567
Gene Name
RAS like estrogen regulated growth inhibitor
Gene Aliases
EGK_03427
Chromosome Location
Chr.11: 16514071 - 16627883 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261674.1 NP_001248603.1 - 414 69
GenBank mRNA JU319806.1 - - 414 69

View Full Product Details