Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRG1
Entrez Gene ID
716727
Gene Name
killer cell lectin like receptor G1
Gene Aliases
-
Chromosome Location
Chr.11: 10745613 - 10767161 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001114131.4 XP_001114131.1 - 286 100

View Full Product Details