Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
706214
Gene Name
matrix remodeling associated 5
Gene Aliases
-
Chromosome Location
Chr.X: 2910742 - 2945734 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086428.4 XP_001086428.3 - 7517 151
GenBank mRNA JU484019.1 - - 2540 151

Gene Symbol
LOC106995402
Entrez Gene ID
106995402
Gene Name
matrix-remodeling-associated protein 5
Gene Aliases
ADLICANY
Chromosome Location
Chr.Y: 1583138 - 1645841 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015128616.2 XP_014984102.2 - 7158 151

View Full Product Details