Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC1A
Entrez Gene ID
717526
Gene Name
C-type lectin domain family 1 member A
Gene Aliases
-
Chromosome Location
Chr.11: 9613253 - 9642616 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266551.1 NP_001253480.1 - 303 149
GenBank mRNA JU475869.1 - - 303 149

View Full Product Details