Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
703497
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 110821116 - 110906012 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139055.2 XP_014994541.2 - 3002 133

View Full Product Details