Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP41
Entrez Gene ID
703215
Gene Name
centrosomal protein 41
Gene Aliases
EGK_14139, TSGA14
Chromosome Location
Chr.3: 156689368 - 156741091 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266838.1 NP_001253767.1 - 694 117
XM_015134895.2 XP_014990381.1 - 945 117
XM_015134896.2 XP_014990382.1 - 701 117
XM_015134898.2 XP_014990384.1 - 2907 117
XM_015134899.2 XP_014990385.1 - 714 117
XM_015134901.2 XP_014990387.1 - 565 117
XM_028844884.1 XP_028700717.1 - 563 117
GenBank mRNA JU321044.1 - - 694 117

View Full Product Details