Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL22RA1
Entrez Gene ID
719264
Gene Name
interleukin 22 receptor subunit alpha 1
Gene Aliases
-
Chromosome Location
Chr.1: 200593789 - 200614993 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015131220.2 XP_014986706.2 - 969 67
XM_015131229.2 XP_014986715.2 - 962 67

View Full Product Details