Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP4K2C
Entrez Gene ID
715120
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Gene Aliases
EGK_03844
Chromosome Location
Chr.11: 57168186 - 57180633 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261154.1 NP_001248083.1 - 589 99
GenBank mRNA JU336433.1 - - 581 99
JU477249.1 - - 558 99
JV048280.1 - - 589 99
JV602242.1 - - 638 99

View Full Product Details