Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIP4K2C
Entrez Gene ID
715120
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
Gene Aliases
EGK_03844
Chromosome Location
Chr.11: 57168186 - 57180633 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261154.1 NP_001248083.1 - 1265 66
GenBank mRNA JU336433.1 - - 1257 66
JU477249.1 - - 1234 66
JV048280.1 - - 1265 66
JV602242.1 - - 1314 66

View Full Product Details