Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXPE3
Entrez Gene ID
702612
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
EGK_11249, FAM55C
Chromosome Location
Chr.2: 174323787 - 174365577 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194109.2 NP_001181038.1 - 869 82
XM_015128755.2 XP_014984241.1 - 1098 82
XM_015128760.2 XP_014984246.1 - 1064 82
XM_015128764.2 XP_014984250.1 - 696 82
XM_028843194.1 XP_028699027.1 - 875 82
GenBank mRNA JU337073.1 - - 687 82
JU337074.1 - - 848 82
JU477804.1 - - 541 82
JV048672.1 - - 318 82

View Full Product Details