Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NXPE3
Entrez Gene ID
702612
Gene Name
neurexophilin and PC-esterase domain family member 3
Gene Aliases
EGK_11249, FAM55C
Chromosome Location
Chr.2: 174323787 - 174365577 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194109.2 NP_001181038.1 - 1907 117
XM_015128755.2 XP_014984241.1 - 2136 117
XM_015128760.2 XP_014984246.1 - 2102 117
XM_015128764.2 XP_014984250.1 - 1734 117
XM_015128769.2 XP_014984255.1 - 1859 117
XM_028843194.1 XP_028699027.1 - 1913 117
GenBank mRNA JU337073.1 - - 1725 117
JU337074.1 - - 1886 117
JU477804.1 - - 1579 117
JV048672.1 - - 1356 117

View Full Product Details