Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGPP2
Entrez Gene ID
705674
Gene Name
sphingosine-1-phosphate phosphatase 2
Gene Aliases
-
Chromosome Location
Chr.12: 109866182 - 110004395 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107876.3 XP_001107876.1 - 648 80
XM_028831148.1 XP_028686981.1 - 209 80
GenBank mRNA JV707003.1 - - 555 80

View Full Product Details