Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9C2
Entrez Gene ID
706596
Gene Name
solute carrier family 9 member C2 (putative)
Gene Aliases
SLC9A10
Chromosome Location
Chr.1: 30789717 - 30891338 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015123900.2 XP_014979386.1 - 1881 84
XM_015123909.2 XP_014979395.1 - 2022 84
XM_015123916.2 XP_014979402.1 - 1928 84
XM_015123920.2 XP_014979406.1 - 1788 84
XM_028850547.1 XP_028706380.1 - 2013 84

View Full Product Details