Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9C1
Entrez Gene ID
708000
Gene Name
solute carrier family 9 member C1
Gene Aliases
SLC9A10
Chromosome Location
Chr.2: 163895852 - 164032239 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015129854.2 XP_014985340.2 - 2627 155
XM_015129855.2 XP_014985341.2 - 2627 155
XM_015129857.2 XP_014985343.2 - 2627 155
XM_015129858.2 XP_014985344.2 - 2627 155
XM_015129859.2 XP_014985345.2 - 2465 155
XM_015129862.2 XP_014985348.1 - 2017 155

View Full Product Details