Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AZGP1
Entrez Gene ID
710136
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Gene Aliases
-
Chromosome Location
Chr.3: 42137134 - 42146412 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194114.2 NP_001181043.2 - 707 118

View Full Product Details