Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLPI
Entrez Gene ID
711156
Gene Name
secretory leukocyte peptidase inhibitor
Gene Aliases
-
Chromosome Location
Chr.10: 80480418 - 80482735 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168663.1 NP_001162134.1 - 102 138

View Full Product Details