Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASTKD5
Entrez Gene ID
100498658
Gene Name
FAST kinase domains 5
Gene Aliases
-
Chromosome Location
Chr.10: 63344252 - 63359440 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190877.2 NP_001177806.2 - 2053 106
XM_015149336.2 XP_015004822.2 - 2371 106
XM_028827103.1 XP_028682936.1 - 2267 106
GenBank mRNA DR768890.1 - - 449 106
JU323380.1 - - 2342 106
JU472405.1 - - 2373 106
JV589888.1 - - 2042 106

View Full Product Details