Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTYMK
Entrez Gene ID
701781
Gene Name
deoxythymidylate kinase
Gene Aliases
-
Chromosome Location
Chr.12: 129103572 - 129115141 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194775.2 NP_001181704.1 - 273 99
XM_001094132.4 XP_001094132.2 - 443 99
XM_015111561.2 XP_014967047.1 - 276 99
GenBank mRNA JV053794.1 - - 182 99

View Full Product Details