Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Etv4
Entrez Gene ID
360635
Gene Name
ets variant 4
Gene Aliases
Pea3
Chromosome Location
Chr.10: 89685058 - 89700314 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108299.1 NP_001101769.1 2 - 3 332 54
XM_008768085.1 XP_008766307.1 2 - 3 402 54

View Full Product Details