The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133528.1 | NP_598212.1 | 1 - 2 | 67 | 130 |
GenBank mRNA | AJ132008.1 | - | 1 - 2 | 67 | 130 |
BC085708.1 | - | 1 - 2 | 88 | 130 | |
FQ229796.1 | - | 1 - 2 | 84 | 129 | |
FQ233766.1 | - | 1 - 2 | 90 | 130 |