Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gata2
Entrez Gene ID
25159
Gene Name
GATA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 120133713 - 120142488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033442.1 NP_254277.1 6 - 7 1583 66
GenBank mRNA AB102789.1 - 4 - 5 1140 66
AB102790.1 - 4 - 5 1140 66
AF345897.1 - 4 - 5 1140 66
AY032734.1 - 6 - 7 1583 66
BC061745.1 - 5 - 6 1390 66
FQ212304.1 - - 1464 66

View Full Product Details