Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnoc
Entrez Gene ID
25516
Gene Name
prepronociceptin
Gene Aliases
N23K, Npnc1
Chromosome Location
Chr.15: 48805842 - 48833053 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013007.1 NP_037139.1 2 - 3 612 100
XM_006252199.2 XP_006252261.1 3 - 4 772 100
GenBank mRNA FQ214340.1 - 3 - 4 659 100
FQ220805.1 - 3 - 4 661 100
S79730.1 - 2 - 3 549 100
U48262.1 - 2 - 3 615 100
X97375.1 - 2 - 3 612 100

View Full Product Details