The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017066.2 | NP_058762.1 | 4 - 5 | 520 | 76 |
XM_006236290.2 | XP_006236352.1 | 4 - 5 | 1904 | 76 | |
XM_006236294.2 | XP_006236356.1 | 4 - 5 | 1500 | 76 | |
GenBank mRNA | BC062013.1 | - | 4 - 5 | 520 | 76 |
FQ213442.1 | - | 4 - 5 | 524 | 76 | |
FQ223195.1 | - | 4 - 5 | 672 | 76 | |
M55601.1 | - | 4 - 5 | 721 | 76 |