The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017238.1 | NP_058934.1 | 1 - 2 | 163 | 57 |
GenBank mRNA | U09631.1 | - | 1 - 2 | 163 | 57 |
Z25885.1 | - | 1 - 2 | 116 | 57 |