The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020074.2 | NP_064459.1 | 1 - 2 | 102 | 74 |
XM_006256483.2 | XP_006256545.1 | 4 - 5 | 482 | 74 | |
XM_006256484.2 | XP_006256546.1 | 2 - 3 | 290 | 74 | |
GenBank mRNA | BC088144.1 | - | 1 - 2 | 102 | 74 |
FQ220127.1 | - | 1 - 2 | 121 | 74 | |
FQ227535.1 | - | 1 - 2 | 119 | 74 | |
FQ231972.1 | - | 1 - 2 | 119 | 74 | |
J03224.1 | - | 1 - 2 | 115 | 74 | |
K02934.1 | - | 1 - 2 | 117 | 74 |