Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Milr1
Entrez Gene ID
59105
Gene Name
mast cell immunoglobulin-like receptor 1
Gene Aliases
Mca32
Chromosome Location
Chr.10: 94944267 - 94961791 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021585.1 NP_067596.1 4 - 5 610 146
GenBank mRNA U39546.1 - 4 - 5 610 146

View Full Product Details