The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031503.2 | NP_113691.1 | 2 - 3 | 1068 | 106 |
GenBank mRNA | X53724.1 | - | 2 - 3 | 1012 | 106 |