The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138908.2 | NP_620263.1 | 7 - 8 | 1365 | 106 |
GenBank mRNA | Y09945.1 | - | 7 - 8 | 1367 | 106 |