Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rmt1
Entrez Gene ID
246306
Gene Name
mammary cancer associated protein RMT-1
Gene Aliases
-
Chromosome Location
Chr.1: 218369204 - 218374619 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145088.1 NP_659556.1 4 - 5 679 133
GenBank mRNA AF308611.1 - 4 - 5 679 133

View Full Product Details