The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145096.2 | NP_659564.2 | 12 - 13 | 1358 | 117 |
XM_008771288.1 | XP_008769510.1 | 11 - 12 | 1295 | 117 | |
GenBank mRNA | AB217870.1 | - | 12 - 13 | 1157 | 117 |
AF228917.1 | - | 12 - 13 | 1347 | 117 | |
AY886521.1 | - | 12 - 13 | 1358 | 117 |