The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173128.1 | NP_775151.1 | 1 - 2 | 416 | 67 |
GenBank mRNA | M63665.1 | - | 1 - 2 | 416 | 67 |