Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdc1
Entrez Gene ID
25216
Gene Name
syndecan 1
Gene Aliases
HSPG, SYNDECA, Synd1, Syndecan
Chromosome Location
Chr.6: 33885576 - 33908038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013026.2 NP_037158.1 3 - 4 844 82
GenBank mRNA BC061834.1 - 3 - 4 844 82
M81785.1 - 3 - 4 859 82
S61865.1 - 3 - 4 859 82
X60651.1 - 2 - 3 534 82

View Full Product Details