Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Entpd2
Entrez Gene ID
64467
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
Cd39l1, NTPDase2
Chromosome Location
Chr.3: 2617795 - 2623818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172030.1 NP_742027.1 3 - 4 389 89
XM_006233644.2 XP_006233706.1 3 - 4 489 89
XM_008761606.1 XP_008759828.1 3 - 4 489 89
GenBank mRNA AF276940.1 - 3 - 4 389 89
BC086558.1 - 3 - 4 399 89

View Full Product Details